Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexadecane

Catalog No. H6703500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
6 x 500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H6703500ML 500 mL
H67036X 6 x 500 mL
H67031L 1 L
H6703100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. H6703500ML Supplier Sigma Aldrich Supplier No. H6703500ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexadecane

Specifications

CAS 544-76-3
Melting Point 18°C
Density 0.773 g/mL at 25°C
Boiling Point 287°C
Molecular Formula C16H34
Linear Formula CH3(CH2)14CH3
Refractive Index n20/D 1.434
MDL Number MFCD00008998
Quantity 500 mL
Synonym n-Hexadecane; Cetane
Molecular Weight (g/mol) 226.44 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.