Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexadecane

Catalog No. H67036X Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
6 x 500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H67036X 6 x 500 mL
H6703500ML 500 mL
H67031L 1 L
H6703100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. H67036X Supplier Sigma Aldrich Supplier No. H67036X500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexadecane

Specifications

CAS 544-76-3
Melting Point 18°C
Density 0.773 g/mL at 25°C
Boiling Point 287°C
Molecular Formula C16H34
Refractive Index n20/D 1.434
Linear Formula CH3(CH2)14CH3
Quantity 6 x 500 mL
MDL Number MFCD00008998
Synonym n-Hexadecane; Cetane
Molecular Weight (g/mol) 226.44 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.