Learn More
Medchemexpress LLC Hexadecane-d34 50mg
SDP
Catalog No. 5000206192
Encompass_Preferred
Hexadecane-d34 (n-Hexadecane-d34) is the deuterium labeled Hexadecane (HY-W099580) Hexadecane (n-Hexadecane) is a saturated hydrocarbon of alkanes or paraffins and can be used as an organic solvent Hexadecane is a component of petroleum belongs to hydrophobic substrates and is a model substance for studying bacterial degradation of hydrophobic compounds[1]
Catalog No. 50-002-06192 Supplier Medchemexpress LLC Supplier No. HYW099580S150MG
Hexadecane-d34 (n-Hexadecane-d34) is the deuterium labeled Hexadecane (HY-W099580) Hexadecane (n-Hexadecane) is a saturated hydrocarbon of alkanes or paraffins and can be used as an organic solvent Hexadecane is a component of petroleum belongs to hydrophobic substrates and is a model substance for studying bacterial degradation of hydrophobic compounds[1]