Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecanedioic acid, 97%, Thermo Scientific™

Catalog No. AC175910025
Change view
Click to view available options
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC175910025 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC175910025 Supplier Thermo Scientific Chemicals Supplier No. 175910025

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 505-54-4
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C16H30O4
Linear Formula HO2C(CH2)14CO2H
Quantity 2.5g
Beilstein 02, 733
Synonym thapsic acid, 1,14-tetradecanedicarboxylic acid, 1,16-hexadecanedioic acid, hexadecane-1,16-dioic acid, 1,14-tetradecane dicarboxylic acid, n-tetradecane-.omega.,.omega.'-dicarboxylic acid, dicarboxylic acid c16, 1,16-hexadecanedioate, thapsate, thaspic acid
InChI Key QQHJDPROMQRDLA-UHFFFAOYSA-N
SMILES C(CCCCCCCC(=O)O)CCCCCCC(=O)O
IUPAC Name hexadecanedioic acid
Molecular Weight (g/mol) 286.412
PubChem CID 10459
ChEBI CHEBI:73722
Formula Weight 286.4
Percent Purity ≥96.0%
Chemical Name or Material Hexadecanedioic acid, 97%
Show More Show Less
EINECSNumber 208-013-5
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.