Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexadecanoate-13C16 potassium | 1458714-74-3 | 98.4% | 310.40 g/mol | 13C16H31KO2 | 1 MG
SDP

Catalog No. 5000283410
Encompass_Preferred

Hexadecanoate-13C16 potassium is a uniformly 13C-labeled hexadecanoate (palmitate) potassium salt supplied as a high-purity analytical standard and tracer for lipid analysis and mass spectrometry applications.

  • Uniformly 13C-labeled at 16 carbons.
  • Potassium salt form.
  • High purity: 98.4%.
  • Low-mass packaging: 1 mg suitable for analytical use.
  • Suitable as an internal standard for mass spectrometry and lipidomics workflows.
  • Molecular formula 13C16H31KO2; molecular weight 310.40 g/mol.

Catalog No. 50-002-83410 Supplier Medchemexpress LLC Supplier No. HYW134007S11MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexadecanoate-13C16 potassium is a uniformly 13C-labeled hexadecanoate (palmitate) potassium salt supplied as a high-purity analytical standard and tracer for lipid analysis and mass spectrometry applications.

  • Uniformly 13C-labeled at 16 carbons.
  • Potassium salt form.
  • High purity: 98.4%.
  • Low-mass packaging: 1 mg suitable for analytical use.
  • Suitable as an internal standard for mass spectrometry and lipidomics workflows.
  • Molecular formula 13C16H31KO2; molecular weight 310.40 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.