Learn More
Medchemexpress LLC Hexadecanoate-13C16 potassium | 1458714-74-3 | 99.0% | 310.40 g/mol | 13C16H31KO2 | 5 MG
SDP
Catalog No. 5000283384
Encompass_Preferred
Hexadecanoate-13C16 potassium is the potassium salt of palmitic (hexadecanoic) acid uniformly labeled with 13C on all 16 carbons. It is supplied as an isotopically labeled standard for use as a tracer or internal standard in analytical chemistry and for biochemical studies involving fatty-acid pathways and stress responses. Reported properties include solubility in ethanol and recommended cold storage conditions.
  • Isotopically labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Potassium salt form compatible with biochemical assays.
  • Soluble in ethanol at approximately 10 mg/mL; ultrasonic assistance may be required.
  • Store sealed at 4°C; in solution store at -80°C up to 6 months.
  • Reported to induce GRP78 and CHOP expression in mouse granulosa cells.
Catalog No. 50-002-83384 Supplier Medchemexpress LLC Supplier No. HYW134007S15MG
Hexadecanoate-13C16 potassium is the potassium salt of palmitic (hexadecanoic) acid uniformly labeled with 13C on all 16 carbons. It is supplied as an isotopically labeled standard for use as a tracer or internal standard in analytical chemistry and for biochemical studies involving fatty-acid pathways and stress responses. Reported properties include solubility in ethanol and recommended cold storage conditions.
  • Isotopically labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Potassium salt form compatible with biochemical assays.
  • Soluble in ethanol at approximately 10 mg/mL; ultrasonic assistance may be required.
  • Store sealed at 4°C; in solution store at -80°C up to 6 months.
  • Reported to induce GRP78 and CHOP expression in mouse granulosa cells.