Learn More
Medchemexpress LLC Hexadecanoate-13C16 potassium | 1458714-74-3 | MFCD04118185 | 99.0% | 310.40 g/mol | 13C16H31KO2 | 10 MG
SDP
Catalog No. 5000283282
Encompass_Preferred
Hexadecanoate-13C16 potassium is the fully 13C-labeled potassium salt of hexadecanoic (palmitic) acid, supplied as an isotopically labeled reagent for research and analytical use. It has molecular formula 13C16H31KO2 and molecular weight 310.40 g/mol and is intended for use as an internal standard or tracer in mass spectrometry and metabolic studies.
  • Fully 13C-labeled potassium salt (13C16).
  • Suitable as a stable-isotope internal standard or tracer.
  • Available in small research quantities, e.g., 10 MG packaging.
  • Purity 99.0%.
  • Store sealed at 4°C, protected from moisture and light.
  • In solvent, store at -80°C for up to 6 months or at -20°C for up to 1 month.
Catalog No. 50-002-83282 Supplier Medchemexpress LLC Supplier No. HYW134007S110MG
Hexadecanoate-13C16 potassium is the fully 13C-labeled potassium salt of hexadecanoic (palmitic) acid, supplied as an isotopically labeled reagent for research and analytical use. It has molecular formula 13C16H31KO2 and molecular weight 310.40 g/mol and is intended for use as an internal standard or tracer in mass spectrometry and metabolic studies.
  • Fully 13C-labeled potassium salt (13C16).
  • Suitable as a stable-isotope internal standard or tracer.
  • Available in small research quantities, e.g., 10 MG packaging.
  • Purity 99.0%.
  • Store sealed at 4°C, protected from moisture and light.
  • In solvent, store at -80°C for up to 6 months or at -20°C for up to 1 month.