Learn More
Medchemexpress LLC Hexadecanoic-12-d acid | 358730-99-1 | 99.9% | 257.43 | C16H31DO2 | 10 MG
SDP
Catalog No. 5000296204
Encompass_Preferred
Palmitic acid-d1 is a deuterium-labeled palmitic acid supplied as a high-purity stable-isotope standard for metabolic and lipidomics research. It is provided as a solid for preparing standard solutions and for use as an internal standard in mass spectrometry and quantitative assays.
  • Deuterium-labeled (d1) palmitic acid for isotope tracing and internal standard use.
  • High purity (99.90%) minimizes isotopic interference.
  • Molecular formula C16H31DO2 and molecular weight 257.43 g/mol.
  • Available in small research sizes suitable for method development and quantitation.
  • Supplied with certificate of analysis and safety data sheet for quality and handling information.
Catalog No. 50-002-96204 Supplier Medchemexpress LLC Supplier No. HYN0830S1810MG
Palmitic acid-d1 is a deuterium-labeled palmitic acid supplied as a high-purity stable-isotope standard for metabolic and lipidomics research. It is provided as a solid for preparing standard solutions and for use as an internal standard in mass spectrometry and quantitative assays.
  • Deuterium-labeled (d1) palmitic acid for isotope tracing and internal standard use.
  • High purity (99.90%) minimizes isotopic interference.
  • Molecular formula C16H31DO2 and molecular weight 257.43 g/mol.
  • Available in small research sizes suitable for method development and quantitation.
  • Supplied with certificate of analysis and safety data sheet for quality and handling information.