Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexadecanoic acid, 2-oxiranylmethyl ester | 7501-44-2 | MFCD14636325 | 98.0% | 312.49 g/mol | C19H36O3 | 25 MG
SDP

Catalog No. 5000283098
Encompass_Preferred

Glycidyl palmitate is an epoxy-bearing long-chain fatty ester supplied as a research-grade biochemical reagent. It is provided as a high-purity white to off-white solid and is intended for assay development, organic synthesis, and studies requiring an epoxide-functional fatty ester. Handle and store according to supplier instructions and safety data sheets.

  • White to off-white solid, easy to weigh and handle.
  • High purity: 98.0%.
  • Molecular weight approximately 312.49 g/mol.
  • Soluble in DMSO at 100 mg/mL with ultrasonic agitation.
  • Stable when stored as a powder at -20 °C for extended periods.
  • Suitable for assay development, organic synthesis, and epoxide chemistry studies.

Catalog No. 50-002-83098 Supplier Medchemexpress LLC Supplier No. HYW12757825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycidyl palmitate is an epoxy-bearing long-chain fatty ester supplied as a research-grade biochemical reagent. It is provided as a high-purity white to off-white solid and is intended for assay development, organic synthesis, and studies requiring an epoxide-functional fatty ester. Handle and store according to supplier instructions and safety data sheets.

  • White to off-white solid, easy to weigh and handle.
  • High purity: 98.0%.
  • Molecular weight approximately 312.49 g/mol.
  • Soluble in DMSO at 100 mg/mL with ultrasonic agitation.
  • Stable when stored as a powder at -20 °C for extended periods.
  • Suitable for assay development, organic synthesis, and epoxide chemistry studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.