Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexadecanoic acid, ethyl ester | 628-97-7 | 284.48 | 250 G
SDP

Catalog No. 5000389422
Encompass_Preferred

Ethyl palmitate is a CHIKV virus inhibitor with an EC50 value of 0.0068 μM. It can reduce levels of TNF-α, IL-6, and NF-κB in endotoxemic rats, demonstrating anti-inflammatory activity.

  • Purity: 99.09%
  • Appearance: solid below 24°C, liquid above 26°C
  • Color: colorless to light yellow
  • Structure classification: lipid
  • Initial source: plants Compositae Dicerothamnus rhinocerotis, plants Umbelliferae

Catalog No. 50-003-89422 Supplier Medchemexpress LLC Supplier No. 250GHYN2086
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl palmitate is a CHIKV virus inhibitor with an EC50 value of 0.0068 μM. It can reduce levels of TNF-α, IL-6, and NF-κB in endotoxemic rats, demonstrating anti-inflammatory activity.

  • Purity: 99.09%
  • Appearance: solid below 24°C, liquid above 26°C
  • Color: colorless to light yellow
  • Structure classification: lipid
  • Initial source: plants Compositae Dicerothamnus rhinocerotis, plants Umbelliferae

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.