Learn More
Medchemexpress LLC Hexadecanoic acid, methyl ester | 112-39-0 | 270.45 | 1 G
SDP
Catalog No. 5000416011
Encompass_Preferred
Methyl palmitate is a naturally occurring fatty acid ester that acts as a potent inhibitor of IκB phosphorylation. It modulates macrophage activity to down-regulate pro-inflammatory mediators such as TNF-α and nitric oxide (NO), exhibiting anti-inflammatory and antifibrotic effects. It can inhibit LPS-induced Kupffer cells and rat peritoneal macrophages, as well as the phagocytic function of RAW cells. This compound is also antagonistic to muscarinic receptors and provides cardioprotective effects against ischemia/reperfusion injury in vivo. It is highly toxic against adult T. cinnabarinus.
  • Naturally occurring fatty acid ester
  • Potent inhibitor of IκB phosphorylation
  • Modulates macrophage activity
  • Down-regulates pro-inflammatory mediators
  • Anti-inflammatory and antifibrotic effects
  • Antagonistic to muscarinic receptors
  • Cardioprotective effects
Catalog No. 50-004-16011 Supplier Medchemexpress LLC Supplier No. HYN14821G
Methyl palmitate is a naturally occurring fatty acid ester that acts as a potent inhibitor of IκB phosphorylation. It modulates macrophage activity to down-regulate pro-inflammatory mediators such as TNF-α and nitric oxide (NO), exhibiting anti-inflammatory and antifibrotic effects. It can inhibit LPS-induced Kupffer cells and rat peritoneal macrophages, as well as the phagocytic function of RAW cells. This compound is also antagonistic to muscarinic receptors and provides cardioprotective effects against ischemia/reperfusion injury in vivo. It is highly toxic against adult T. cinnabarinus.
  • Naturally occurring fatty acid ester
  • Potent inhibitor of IκB phosphorylation
  • Modulates macrophage activity
  • Down-regulates pro-inflammatory mediators
  • Anti-inflammatory and antifibrotic effects
  • Antagonistic to muscarinic receptors
  • Cardioprotective effects