Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecenylsuccinic Anhydride 94.0+%, TCI America™
SDP

Catalog No. H007325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H007325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H007325G Supplier TCI America Supplier No. H007325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32072-96-1
Molecular Formula C20H34O3
Molecular Weight (g/mol) 322.489
MDL Number MFCD00014544
InChI Key RSPWVGZWUBNLQU-FOCLMDBBSA-N
PubChem CID 5354625
IUPAC Name 3-[(E)-hexadec-1-enyl]oxolane-2,5-dione
SMILES CCCCCCCCCCCCCCC=CC1CC(=O)OC1=O

Specifications

Color White-Yellow
Boiling Point 410°C
Quantity 25 g
Formula Weight 322.49
Percent Purity ≥94.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecenylsuccinic Anhydride
EINECSNumber (2)-0852&(2)-1140
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.