Learn More
Cayman Chemical Hexadecyl Acetyl Glycerol 50mg
SDP
Catalog No. 502894424
Encompass_Preferred
An analog of DAG, which inhibits the activation of PKC by DAG
Catalog No. 50-289-4424 Supplier Cayman Chemical Supplier No. 6092050 MG
An analog of DAG, which inhibits the activation of PKC by DAG