Learn More
Medchemexpress LLC Hexadecyl β-D-maltoside | 98064-96-1 | MFCD01310880 | 99.8% | 566.72 g·mol⁻¹ | C28H54O11 | 10 MG
SDP
Catalog No. 5000301393
Encompass_Preferred
Hexadecyl β-D-maltoside is a nonionic alkyl-maltoside detergent used to solubilize and stabilize membrane proteins for biochemical assays and structural studies. Supplied as a white to off-white solid, it is suitable for preparation of concentrated stock solutions for downstream assays.
  • Nonionic detergent for membrane protein solubilization.
  • Preserves protein activity and native conformation.
  • High purity suitable for biochemical assays.
  • Forms clear solutions at typical working concentrations.
  • Stable when stored as powder at -20°C or 4°C.
  • Available in small research pack sizes (10 mg, 25 mg).
Catalog No. 50-003-01393 Supplier Medchemexpress LLC Supplier No. HYW02079510MG
Hexadecyl β-D-maltoside is a nonionic alkyl-maltoside detergent used to solubilize and stabilize membrane proteins for biochemical assays and structural studies. Supplied as a white to off-white solid, it is suitable for preparation of concentrated stock solutions for downstream assays.
  • Nonionic detergent for membrane protein solubilization.
  • Preserves protein activity and native conformation.
  • High purity suitable for biochemical assays.
  • Forms clear solutions at typical working concentrations.
  • Stable when stored as powder at -20°C or 4°C.
  • Available in small research pack sizes (10 mg, 25 mg).