Learn More
Medchemexpress LLC Hexadecyl palmitate | 540-10-3 | 98.1% | 480.86 | 1 KG
SDP
Catalog No. 5000366329
Encompass_Preferred
Hexadecyl palmitate is an organic compound commonly used in the cosmetic industry as an emollient, thickener, and skin conditioning agent. It is derived from a mixture of cetyl alcohol (a fatty alcohol) and palmitic acid (a saturated fatty acid). It is often used in skin care products such as moisturizers, lotions, and creams to improve their texture and helps them glide smoothly on the skin. In addition to its use in cosmetics, cetyl palmitate is used in other industries including pharmaceuticals, food additives, and lubricants. Overall, it is considered safe for use in personal care products, but like any other ingredient, it may cause irritation or an allergic reaction in some people. Hexadecyl palmitate is also a biochemical reagent that can be used as a biological material or organic compound for life science related research.
Catalog No. 50-003-66329 Supplier Medchemexpress LLC Supplier No. HYW0108071KG
Hexadecyl palmitate is an organic compound commonly used in the cosmetic industry as an emollient, thickener, and skin conditioning agent. It is derived from a mixture of cetyl alcohol (a fatty alcohol) and palmitic acid (a saturated fatty acid). It is often used in skin care products such as moisturizers, lotions, and creams to improve their texture and helps them glide smoothly on the skin. In addition to its use in cosmetics, cetyl palmitate is used in other industries including pharmaceuticals, food additives, and lubricants. Overall, it is considered safe for use in personal care products, but like any other ingredient, it may cause irritation or an allergic reaction in some people. Hexadecyl palmitate is also a biochemical reagent that can be used as a biological material or organic compound for life science related research.