Learn More
Medchemexpress LLC Hexadecyl palmitate | 540-10-3 | 98.1% | 480.86 | 25 G
SDP
Catalog No. 5000451657
Encompass_Preferred
Hexadecyl palmitate is an organic compound and analytical standard used in research and analytical applications. It functions as an emollient, thickener, and skin conditioning agent, primarily in the cosmetic industry for products like moisturizers, lotions, and creams to enhance texture and smooth application. Its applications extend to pharmaceuticals, food additives, and lubricants.
  • Functions as an emollient, thickener, and skin conditioning agent
  • Used in skin care products to improve texture and aid smooth application
  • Applicable in pharmaceuticals, food additives, and lubricants
  • Intended for research and analytical applications
Catalog No. 50-004-51657 Supplier Medchemexpress LLC Supplier No. 25GHYW010807
Hexadecyl palmitate is an organic compound and analytical standard used in research and analytical applications. It functions as an emollient, thickener, and skin conditioning agent, primarily in the cosmetic industry for products like moisturizers, lotions, and creams to enhance texture and smooth application. Its applications extend to pharmaceuticals, food additives, and lubricants.
  • Functions as an emollient, thickener, and skin conditioning agent
  • Used in skin care products to improve texture and aid smooth application
  • Applicable in pharmaceuticals, food additives, and lubricants
  • Intended for research and analytical applications