Learn More
Medchemexpress LLC Hexadecyl palmitate | 540-10-3 | 98.1% | 480.86 | 50 G
SDP
Catalog No. 5000366331
Encompass_Preferred
Hexadecyl palmitate is an organic compound commonly used in the cosmetic industry as an emollient, thickener, and skin conditioning agent. It is derived from a mixture of cetyl alcohol and palmitic acid. It is frequently incorporated into skincare products such as moisturizers, lotions, and creams to enhance their texture and facilitate smooth application on the skin. It is also a biochemical reagent that can be utilized as a biological material or organic compound for life science-related research.
  • Used as an emollient, thickener, and skin conditioning agent
  • Derived from cetyl alcohol and palmitic acid
  • Enhances texture and smooth application in skincare products
  • Applications in pharmaceuticals, food additives, and lubricants
  • Considered safe for personal care products
  • Biochemical reagent for life science research
Catalog No. 50-003-66331 Supplier Medchemexpress LLC Supplier No. HYW01080750G
Hexadecyl palmitate is an organic compound commonly used in the cosmetic industry as an emollient, thickener, and skin conditioning agent. It is derived from a mixture of cetyl alcohol and palmitic acid. It is frequently incorporated into skincare products such as moisturizers, lotions, and creams to enhance their texture and facilitate smooth application on the skin. It is also a biochemical reagent that can be utilized as a biological material or organic compound for life science-related research.
  • Used as an emollient, thickener, and skin conditioning agent
  • Derived from cetyl alcohol and palmitic acid
  • Enhances texture and smooth application in skincare products
  • Applications in pharmaceuticals, food additives, and lubricants
  • Considered safe for personal care products
  • Biochemical reagent for life science research