Learn More
Medchemexpress LLC Hexadecyl palmitate | 540-10-3 | 98.1% | 480.86 | 500 G
SDP
Catalog No. 5000453000
Encompass_Preferred
Hexadecyl palmitate is an organic compound commonly used in the cosmetic industry as an emollient, thickener, and skin conditioning agent. It is derived from a mixture of cetyl alcohol (a fatty alcohol) and palmitic acid (a saturated fatty acid). It is often used in skincare products such as moisturizers, lotions, and creams to improve their texture and help them glide smoothly on the skin. In addition to cosmetics, it is used in pharmaceuticals, food additives, and lubricants. It is considered safe for use in personal care products, but like any other ingredient, it may cause irritation or an allergic reaction in some people.
  • Can be used as a biochemical reagent for life science-related research.
Catalog No. 50-004-53000 Supplier Medchemexpress LLC Supplier No. HYW010807500G
Hexadecyl palmitate is an organic compound commonly used in the cosmetic industry as an emollient, thickener, and skin conditioning agent. It is derived from a mixture of cetyl alcohol (a fatty alcohol) and palmitic acid (a saturated fatty acid). It is often used in skincare products such as moisturizers, lotions, and creams to improve their texture and help them glide smoothly on the skin. In addition to cosmetics, it is used in pharmaceuticals, food additives, and lubricants. It is considered safe for use in personal care products, but like any other ingredient, it may cause irritation or an allergic reaction in some people.
  • Can be used as a biochemical reagent for life science-related research.