Learn More
AdipoGen Hexadecylpyrene-1-sulfonamide
SDP
Catalog No. 502560770
Encompass_Preferred
Chemical. CAS 351002-71-6. Formula C32H43NO2S. MW 505.8. Synthetic. Fluorescent probe. Spectral properties lambdaex=350nm lambdaem=400nm in MeOH. May be used in the analysis of drug delivery devices for lipophilic pyrenyl compounds.
Catalog No. 50-256-0770 Supplier AdipoGen Supplier No. CDXH0070M100
Chemical. CAS 351002-71-6. Formula C32H43NO2S. MW 505.8. Synthetic. Fluorescent probe. Spectral properties lambdaex=350nm lambdaem=400nm in MeOH. May be used in the analysis of drug delivery devices for lipophilic pyrenyl compounds.