Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexadecyltrimethoxysilane

Catalog No. 52360100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-360-100ML 100 mL
52-360-500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52-360-100ML Supplier Sigma Aldrich Supplier No. 52360100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexadecyltrimethoxysilane (HDTMS)-ßmay be used to prepare hydrophobic silica aerogels by sol-gel process.{80} HDTMS was coated upon silica covered aluminum pigments.{81} Metal nanoparticles (CeO2, Fe2O4, TiO2, ZnO) coated with HDTMS form stable dispersions in hydrocarbon solvents. {82} HDTMS may be used to functionalize polyisocyanurate (PIR) foams {84} and graphene.{83}

Specifications

CAS 16415-12-6
Density 0.89 g/mL (at 20°C)
Molecular Formula C19H42O3Si
Refractive Index n20/D 1.44
Linear Formula H3C(CH2)15 Si(OCH3)3
Quantity 100 mL
MDL Number MFCD00069149
Synonym Trimethoxyhexadecylsilane
Molecular Weight (g/mol) 346.62
Percent Purity ≥85% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.