Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecyltrimethylammonium bromide, 98-102%, MilliporeSigma™ Supelco™

Catalog No. 111019509 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9509 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9509 Supplier MilliporeSigma Supelco Supplier No. 369321G
Add to Cart
Add to Cart

Cetrimonium bromide(CTAB) is a quaternary ammonium derivative known to enhance anti-tumor activity. It is one of the components of the broad-spectrum antiseptic cetrimide and may also be used as a buffer for DNA extraction.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 57-09-0
Flash Point 244°C - Closed Cup
Physical Form Neat
Linear Formula CH3(CH2)15 N(Br)(CH3)3
MDL Number MFCD00011772
Melting Point 248°C to 251°C
Synonym CTAB; Cetrimonium bromide; Cetyltrimethylammonium bromide; Palmityltrimethylammonium bromide
Solubility Completely soluble in water
Type Analytical Standard
Quantity 1 g
Formula Weight 364.45
Percent Purity 98 to 102%
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.