Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecyltrimethylammonium Hexafluorophosphate 98.0+%, TCI America™
SDP

Catalog No. H073325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H073325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H073325G Supplier TCI America Supplier No. H073325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 101079-29-2
Melting Point 185°C
Color White
Molecular Formula C19H42F6NP
Quantity 25 g
MDL Number MFCD00144894
Synonym Cetyltrimethylammonium Hexafluorophosphate
InChI Key XIHYIALOMDSESV-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)C.F[P-](F)(F)(F)(F)F
IUPAC Name hexadecyl(trimethyl)azanium;hexafluorophosphate
Molecular Weight (g/mol) 429.516
PubChem CID 23500182
Formula Weight 429.52
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecyltrimethylammonium Hexafluorophosphate
Show More Show Less
EINECSNumber (2)-0184&(9)-0795&(9)-1971
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.