Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecyltrimethylammonium Hydrogen Sulfate 98.0+%, TCI America™
SDP

Catalog No. H161625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H161625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H161625G Supplier TCI America Supplier No. H161625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68214-07-3
Molecular Formula C19H43NO4S
Molecular Weight (g/mol) 381.616
MDL Number MFCD00134393
InChI Key UCRJJNVFJGKYQT-UHFFFAOYSA-M
Synonym cetyltrimethylammonium hydrogensulfate, hexadecyltrimethylammonium hydrogensulphate, 1-hexadecanaminium, n,n,n-trimethyl-, sulfate 1:1, hexadecyltrimethylammonium hydrogen sulfate, cetyltrimethylammonium hydrogen sulfate, n,n,n-trimethylhexadecan-1-aminium hydrogensulfate, hexadecyltrimethylammonium hydrogen sulphate, ammonium, hexadecyltrimethyl-, hydrogen sulfate, hexadecyltrimethylammonium bisulfate, n,n,n-trimethylhexadecan-1-aminium hydrogen sulfate 1:1:1
PubChem CID 109763
IUPAC Name hexadecyl(trimethyl)azanium;hydrogen sulfate
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)C.OS(=O)(=O)[O-]

Specifications

Melting Point 260°C
Color White
Quantity 25 g
Formula Weight 381.62
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecyltrimethylammonium Hydrogen Sulfate
EINECSNumber (2)-0184&(9)-1971
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.