Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexadecyltrimethylammonium Hydroxide Solution

Catalog No. 43923125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-923-125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 43-923-125ML Supplier Sigma Aldrich Supplier No. 43923125ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexadecyltrimethylammonium hydroxide solution is a surfactant used in:Synthesis of microporous and mesoporous aluminosilicate molecular sieves such as MCM-41 and MCM-48.Hydrothermal preparation of manganese dioxide (MnO2) nanoflower.Synthesis of cetyltrimethylammonium hydroperoxide, a reagent used for promoting phosphate ester hydrolysis.

Specifications

CAS 149-74-6
Density 1.176 g/mL at 25°C
Boiling Point 205°C
Molecular Formula C7H8Cl2Si
Refractive Index n20/D 1.351
Linear Formula C6H5Si(CH3)Cl2
Quantity 25 mL
MDL Number MFCD00000490
Molecular Weight (g/mol) 191.13 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.