Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexadecyltrimethylammonium Hydroxide Solution

Catalog No. 5238750ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-387-50ML 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 52-387-50ML Supplier Sigma Aldrich Supplier No. 5238750ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexadecyltrimethylammonium hydroxide solution is a commonly used phase transfer agent and surfactant solution employed in the synthesis of a variety of mesoporous materials, micelle complexes and transition-metal oxides.

Specifications

CAS 180516-87-4
Melting Point 160°C to 165°C
Molecular Formula C20H24Cl2Zr
Refractive Index n20/D 1.372
Linear Formula C20H24Cl2Zr
MDL Number MFCD00672123
Quantity 100 mg
Synonym CTAOH solution; Cetrimonium hydroxide solution; Cetyltrimethylammonium hydroxide
Molecular Weight (g/mol) 426.53 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.