Learn More
Sigma Aldrich Fine Chemicals Biosciences Hexadecyltrimethylammonium bromide, 57-09-0, MFCD00011772, 500 g
Catalog No. NC1188379
Encompass
Linear Formula: CH3(CH2)15N(Br)(CH3)3, Molecular Weight: 364.45, ≥98%, Synonym: CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide.
Catalog No. NC1188379 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H5882500G
Linear Formula: CH3(CH2)15N(Br)(CH3)3, Molecular Weight: 364.45, ≥98%, Synonym: CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide.