Learn More
Medchemexpress LLC Hexaenoic acid-d6 1mg | 2692624-21-6 | 502.84 | C34H50D6O2 | 1 MG
SDP
Catalog No. 5000229433
Encompass_Preferred
Hexaenoic acid-d6 is the deuterium-labeled analog of hexaenoic acid supplied as a colorless oil for use as a stable isotope internal standard in analytical and biochemical research. It provides defined isotopic labeling and high chemical purity to support accurate quantitation by LC-MS and related methods.
  • Deuterium-labeled internal standard for LC-MS quantitation.
  • High chemical and isotopic purity for reliable results.
  • Supplied as an oil for convenient handling and formulation.
  • Recommended storage conditions preserve stability in solvent and neat forms.
  • Suitable for studies of very long chain polyunsaturated fatty acids.
Catalog No. 50-002-29433 Supplier Medchemexpress LLC Supplier No. HY117057S1MG
Hexaenoic acid-d6 is the deuterium-labeled analog of hexaenoic acid supplied as a colorless oil for use as a stable isotope internal standard in analytical and biochemical research. It provides defined isotopic labeling and high chemical purity to support accurate quantitation by LC-MS and related methods.
  • Deuterium-labeled internal standard for LC-MS quantitation.
  • High chemical and isotopic purity for reliable results.
  • Supplied as an oil for convenient handling and formulation.
  • Recommended storage conditions preserve stability in solvent and neat forms.
  • Suitable for studies of very long chain polyunsaturated fatty acids.