Learn More
Medchemexpress LLC Hexaethylene glycol | 2615-15-8 | 99.5% | 282.33 | 25 MG
SDP
Catalog No. 5000402256
Encompass_Preferred
Hexaethylene glycol is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It is intended for research purposes only and not for patient use.
  • Used in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-004-02256 Supplier Medchemexpress LLC Supplier No. 25MGHY29504
Hexaethylene glycol is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It is intended for research purposes only and not for patient use.
  • Used in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only