Learn More
Medchemexpress LLC Hexaethylene glycol | 2615-15-8 | 99.5% | C12H26O7 | 10 G
SDP
Catalog No. 5000356025
Encompass_Preferred
Hexaethylene glycol is a PEG-based PROTAC linker that plays a crucial role in the synthesis of PROTACs. These molecules are designed with two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other binding to a target protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of specific target proteins.
  • Acts as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTAC molecules
  • Facilitates the degradation of target proteins via the ubiquitin-proteasome system
  • For research use only
Catalog No. 50-003-56025 Supplier Medchemexpress LLC Supplier No. HY14123010G
Hexaethylene glycol is a PEG-based PROTAC linker that plays a crucial role in the synthesis of PROTACs. These molecules are designed with two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other binding to a target protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of specific target proteins.
  • Acts as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTAC molecules
  • Facilitates the degradation of target proteins via the ubiquitin-proteasome system
  • For research use only