Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexaethylene glycol | 2615-15-8 | 99.5% | C12H26O7 | 25 G
SDP

Catalog No. 5000357342
Encompass_Preferred

Hexaethylene glycol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two different ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets a specific protein, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.

  • Molecular weight: 282.33
  • Appearance: colorless to light yellow liquid
  • Purity: 99.53%
  • CAS number: 2615-15-8
  • Chemical formula: C12H26O7
  • Melting/freezing point: 5-7 °C
  • Boiling point: 217 °C at 4 mmHg
  • Flash point: >110 °C
  • Identified uses: laboratory chemicals, manufacture of substances
  • Recommended storage (pure form): -20 °C for 3 years; 4 °C for 2 years
  • Recommended storage (in solvent): -80 °C for 6 months; -20 °C for 1 month

Catalog No. 50-003-57342 Supplier Medchemexpress LLC Supplier No. 25GHY141230
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexaethylene glycol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two different ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets a specific protein, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.

  • Molecular weight: 282.33
  • Appearance: colorless to light yellow liquid
  • Purity: 99.53%
  • CAS number: 2615-15-8
  • Chemical formula: C12H26O7
  • Melting/freezing point: 5-7 °C
  • Boiling point: 217 °C at 4 mmHg
  • Flash point: >110 °C
  • Identified uses: laboratory chemicals, manufacture of substances
  • Recommended storage (pure form): -20 °C for 3 years; 4 °C for 2 years
  • Recommended storage (in solvent): -80 °C for 6 months; -20 °C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.