Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexaethylene glycol | 2615-15-8 | 99.5% | C12H26O7 | 5 G
SDP

Catalog No. 5000355303
Encompass_Preferred

Hexaethylene glycol is a PEG-based PROTAC linker, which is a key component in the synthesis of PROTACs. PROTACs function by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Serves as a linker in the synthesis of PROTACs.
  • Polyethylene glycol (PEG)-based compound.
  • Intended for research purposes only, not for human use.
  • Purity of 99.5%.
  • Colorless to light yellow liquid.
  • Molecular weight: 282.33.

Catalog No. 50-003-55303 Supplier Medchemexpress LLC Supplier No. HY1412305G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexaethylene glycol is a PEG-based PROTAC linker, which is a key component in the synthesis of PROTACs. PROTACs function by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Serves as a linker in the synthesis of PROTACs.
  • Polyethylene glycol (PEG)-based compound.
  • Intended for research purposes only, not for human use.
  • Purity of 99.5%.
  • Colorless to light yellow liquid.
  • Molecular weight: 282.33.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.