Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexaethylene glycol monobenzyl ether | 24342-68-5 | MFCD06797107 | 99.9% | 372.45 | C19H32O7 | 1 G
SDP

Catalog No. 5000203154
Encompass_Preferred

BnO-PEG6-OH (hexaethylene glycol monobenzyl ether) is a benzyl-protected, six-unit polyethylene glycol linker used as a short hydrophilic spacer in bioconjugation chemistry. It is commonly used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based linker in PROTAC design.

  • Cas number: 24342-68-5.
  • Purity: 99.9% (as supplied).
  • Molecular formula: C19H32O7.
  • Molecular weight: 372.45.
  • Typical applications: ADC linker synthesis and PROTAC linker synthesis.
  • Available pack sizes: 250 mg, 500 mg, 1 g, 5 g, 10 g, 25 g, 50 g.

Catalog No. 50-002-03154 Supplier Medchemexpress LLC Supplier No. HYW0426541G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BnO-PEG6-OH (hexaethylene glycol monobenzyl ether) is a benzyl-protected, six-unit polyethylene glycol linker used as a short hydrophilic spacer in bioconjugation chemistry. It is commonly used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based linker in PROTAC design.

  • Cas number: 24342-68-5.
  • Purity: 99.9% (as supplied).
  • Molecular formula: C19H32O7.
  • Molecular weight: 372.45.
  • Typical applications: ADC linker synthesis and PROTAC linker synthesis.
  • Available pack sizes: 250 mg, 500 mg, 1 g, 5 g, 10 g, 25 g, 50 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.