Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexaethylene glycol phosphoramidite | 125607-09-2 | C42H61N2O10P | 100 MG
SDP

Catalog No. 5000372054
Encompass_Preferred

Hexaethylene glycol phosphoramidite, also known as Spacer Phosphoramidite 18, is an amidite reagent used for oligonucleotide synthesis. It functions as a linker in the synthesis of nucleotide chains and qPCR probes, offering a purity of 99.26%.

  • Amidite reagent for oligonucleotide synthesis
  • Functions as a linker in nucleotide chain creation
  • Utilized in qPCR probe synthesis
  • Appearance: liquid, colorless to light yellow
  • Soluble in DMSO (50 mg/mL, requires ultrasonication)

Catalog No. 50-003-72054 Supplier Medchemexpress LLC Supplier No. HY151651100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexaethylene glycol phosphoramidite, also known as Spacer Phosphoramidite 18, is an amidite reagent used for oligonucleotide synthesis. It functions as a linker in the synthesis of nucleotide chains and qPCR probes, offering a purity of 99.26%.

  • Amidite reagent for oligonucleotide synthesis
  • Functions as a linker in nucleotide chain creation
  • Utilized in qPCR probe synthesis
  • Appearance: liquid, colorless to light yellow
  • Soluble in DMSO (50 mg/mL, requires ultrasonication)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.