Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexaethylene glycol phosphoramidite | 125607-09-2 | C42H61N2O10P | 25 MG
SDP

Catalog No. 5000375995
Encompass_Preferred

Hexaethylene glycol phosphoramidite, also known as Spacer Phosphoramidite 18, is an amidite reagent designed for oligonucleotide synthesis. This compound also serves as a versatile linker in the creation of nucleotide chains and qPCR probes.

  • Functions as an amidite reagent for oligonucleotide synthesis
  • Can be used as a linker in nucleotide chain synthesis
  • Can be used as a linker in qPCR probe synthesis
  • Purity of 99.26%
  • Appears as a liquid, colorless to light yellow
  • Soluble in DMSO at 50 mg/mL
  • Recommended storage for pure form: -20°C for 3 years or 4°C for 2 years
  • Recommended storage for in-solvent form: -80°C for 6 months or -20°C for 1 month
  • For research use only

Catalog No. 50-003-75995 Supplier Medchemexpress LLC Supplier No. 25MGHY151651
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexaethylene glycol phosphoramidite, also known as Spacer Phosphoramidite 18, is an amidite reagent designed for oligonucleotide synthesis. This compound also serves as a versatile linker in the creation of nucleotide chains and qPCR probes.

  • Functions as an amidite reagent for oligonucleotide synthesis
  • Can be used as a linker in nucleotide chain synthesis
  • Can be used as a linker in qPCR probe synthesis
  • Purity of 99.26%
  • Appears as a liquid, colorless to light yellow
  • Soluble in DMSO at 50 mg/mL
  • Recommended storage for pure form: -20°C for 3 years or 4°C for 2 years
  • Recommended storage for in-solvent form: -80°C for 6 months or -20°C for 1 month
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.