Learn More
Cayman Chemical Hexaflumuron, Purity: ≥95%, CAS: 86479-06-3, 50 mg
SDP
Catalog No. 502965644
Encompass_Preferred
A chitin synthesis inhibitor that is used to bait and eliminate termite colonies; also effective against the raisin moth, cowpea weevil, and Asiatic rice borer
Catalog No. 50-296-5644 Supplier Cayman Chemical Supplier No. 1859850 MG
A chitin synthesis inhibitor that is used to bait and eliminate termite colonies; also effective against the raisin moth, cowpea weevil, and Asiatic rice borer