Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexafluoro-2,2-diphenylpropane 98.0+%, TCI America™
SDP

Catalog No. H09785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H09785G Supplier TCI America Supplier No. H09785G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 83558-76-3
Molecular Formula C15H10F6
Molecular Weight (g/mol) 304.235
MDL Number MFCD02093476
InChI Key CFTSORNHIUMCGF-UHFFFAOYSA-N
Synonym (Hexafluoroisopropylidene)bisbenzene
PubChem CID 14047936
IUPAC Name (1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-yl)benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(F)(F)F)C(F)(F)F

Specifications

Color Colorless
Quantity 5 g
Formula Weight 304.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexafluoro-2,2-diphenylpropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.