Learn More
Medchemexpress LLC Hexafluoro-diallylhexaflurobisphenol | 128481-73-2 | 97.4% | 5 MG
SDP
Catalog No. 5000232790
Encompass_Preferred
Hexafluoro-diallylhexaflurobisphenol is a synthetic Honokiol analogue used in biochemical and pharmacological research. It is a hexafluoro-substituted bisphenol derivative supplied as a high-purity oil for in vitro assays and medicinal chemistry studies.
  • Used as a Honokiol analogue for antioxidative and anticancer research.
  • High purity suitable for analytical and screening applications.
  • Small milligram pack sizes support early-stage assay work.
  • Molecular weight 416.36 g/mol and formula C21H18F6O2.
  • Oil form facilitates dissolution in organic solvents.
Catalog No. 50-002-32790 Supplier Medchemexpress LLC Supplier No. HY1207845MG
Hexafluoro-diallylhexaflurobisphenol is a synthetic Honokiol analogue used in biochemical and pharmacological research. It is a hexafluoro-substituted bisphenol derivative supplied as a high-purity oil for in vitro assays and medicinal chemistry studies.
  • Used as a Honokiol analogue for antioxidative and anticancer research.
  • High purity suitable for analytical and screening applications.
  • Small milligram pack sizes support early-stage assay work.
  • Molecular weight 416.36 g/mol and formula C21H18F6O2.
  • Oil form facilitates dissolution in organic solvents.