Learn More
Strem, An Ascensus Company CAS# 392-56-3. 5g. Hexafluorobenzene, min. 99%. MFCD00000288
SDP
Catalog No. 5090109339
Encompass
CAS# 392-56-3. 5g. Hexafluorobenzene, min. 99%. MFCD00000288. Molecular Weight: 186.05. Molecular Formula: C6F6. Color/form: colorless liq. Strem# 09-4630. http://www.strem.com/catalog/v/09-4630/
Catalog No. 50-901-09339 Supplier Strem, An Ascensus Company Supplier No. 0946305G
CAS# 392-56-3. 5g. Hexafluorobenzene, min. 99%. MFCD00000288. Molecular Weight: 186.05. Molecular Formula: C6F6. Color/form: colorless liq. Strem# 09-4630. http://www.strem.com/catalog/v/09-4630/