Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexafluoroglutaric Acid 97.0+%, TCI America™
SDP

Catalog No. H065825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H065825G 25 g
H065810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H065825G Supplier TCI America Supplier No. H065825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 376-73-8
Molecular Formula C5H2F6O4
Molecular Weight (g/mol) 240.06
MDL Number MFCD00004172
InChI Key CCUWGJDGLACFQT-UHFFFAOYSA-N
Synonym hexafluoroglutaric acid, perfluoroglutaric acid, hexafluoropentanedioic acid, pentanedioic acid, hexafluoro, glutaric acid, hexafluoro, pentanedioic acid, 2,2,3,3,4,4-hexafluoro, acmc-209iut, perfluoropentanedioic acid, glutaric acid, hexafluoro-8ci
PubChem CID 67827
IUPAC Name hexafluoropentanedioic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(O)=O

Specifications

Melting Point 97°C
Color Red-White
Quantity 25 g
Formula Weight 240.06
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hexafluoroglutaric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.