Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexafluorophosphoric acid, ca 60% w/w aq. soln.

Catalog No. AAL15728AP
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15728AP 500 mL
AAL15728AC 25 mL
AAL15728AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL15728AP Supplier Thermo Scientific Chemicals Supplier No. L15728AP
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16940-81-1 | HPF6

Hexafluorophosphoric acid is used as a catalyst. It is also used as a metal cleaner and chemical polishing agent for the formation of protective coatings of metal surfaces..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexafluorophosphoric acid is used as a catalyst. It is also used as a metal cleaner and chemical polishing agent for the formation of protective coatings of metal surfaces..

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong bases, glass, alkali metals, powdered metals, strong oxidizing agents, metals and reacts violently with water.  
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16940-81-1
Molecular Formula HPF6
MDL Number MFCD00011350

Specifications

Density 1.65
Quantity 500 mL
UN Number UN1782
Solubility Information Immiscible with water.
Formula Weight 145.97
Concentration or Composition (by Analyte or Components) ≈60% w/w aq. soln.
Chemical Name or Material Hexafluorophosphoric acid
Hazard Category H300+H310+H330-H314-H335
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HEXAFLUOROPHOSPHORIC ACID
EINECSNumber 241-006-5
RTECSNumber SY7115000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.