Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexagonal boron nitride ink

Catalog No. 9014105ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-141-05ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-141-05ML Supplier Sigma Aldrich Supplier No. 9014105ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexagonal boron nitride ink (hBN) is an insulating material with properties like high thermal conductivity, excellent dimensional stability, and good mechanical robustness. hBN inks can be used in the fabrication of flexible memory devices.Sold under Materials Transfer Agreement with Mark Hersam group at Northwestern University.

Specifications

Boiling Point 213°C to 218°C (Terpineol)
CAS 10043-11-5
Quantity 5 mL
Molecular Formula BN
Linear Formula BN
MDL Number MFCD00011317
Synonym Hexagonal boron nitride dispersion, with ethyl cellulose in cyclohexanone and terpineol; hBN dispersion; hBN inkjet printable ink
Molecular Weight (g/mol) 24.82
Density 0.9-1.1 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.