Learn More
Electron Microscopy Sciences Veco Hexagonal 100 mesh Copper Grid
SDP
Catalog No. 5026489
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter, 0.8 mil thickness.
Grid: Copper
Pitch: 250 µm
Hole: 200 µm
Bar: 50 µm
Catalog No. 50-264-89 Supplier Electron Microscopy Sciences Supplier No. H100CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter, 0.8 mil thickness.
Grid: Copper
Pitch: 250 µm
Hole: 200 µm
Bar: 50 µm