Learn More
Electron Microscopy Sciences Veco Hexagonal 150 mesh Nickel Grid
SDP
Catalog No. 5026483
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter, 0.8 mil thickness.
Grid: Nickel
Pitch: 167 µm
Hole: 117 µm
Bar: 50 µm
Catalog No. 50-264-83 Supplier Electron Microscopy Sciences Supplier No. H150NI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter, 0.8 mil thickness.
Grid: Nickel
Pitch: 167 µm
Hole: 117 µm
Bar: 50 µm