Learn More
Electron Microscopy Sciences Gilder Hexagonal 300  mesh Gold Grid
SDP
Catalog No. 5029059
Encompass_Preferred
A reliable support specimen grid source. Features: well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. The copper grids with palladium plating offers better grid strength and avoids tarnishing. 3.05 mm diameter, 0.7 mil (18um) thickness.
Grid: Gold
Pitch: 83 µm
Hole: 58 µm
Bar: 25 µm
Catalog No. 50-290-59 Supplier Electron Microscopy Sciences Supplier No. G300HAU
A reliable support specimen grid source. Features: well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. The copper grids with palladium plating offers better grid strength and avoids tarnishing. 3.05 mm diameter, 0.7 mil (18um) thickness.
Grid: Gold
Pitch: 83 µm
Hole: 58 µm
Bar: 25 µm