Learn More
Electron Microscopy Sciences Veco Hexagonal H111KSpec Copper Grid
SDP
Catalog No. 5026487
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter, 0.8 mil thickness.
Grid: Copper
Hole: 185 µm
Catalog No. 50-264-87 Supplier Electron Microscopy Sciences Supplier No. H111KCU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter, 0.8 mil thickness.
Grid: Copper
Hole: 185 µm