Learn More
Electron Microscopy Sciences Gilder Hexagonal 100 mesh Nickel Grid
SDP
Catalog No. 5029053
Encompass_Preferred
A reliable support specimen grid source. Features: well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. The copper grids with palladium plating offers better grid strength and avoids tarnishing. 3.05 mm diameter, 0.7 mil (18um) thickness.
Grid: Nickel
Pitch: 250 µm
Hole: 215 µm
Bar: 35 µm
Catalog No. 50-290-53 Supplier Electron Microscopy Sciences Supplier No. G100HNI
A reliable support specimen grid source. Features: well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. The copper grids with palladium plating offers better grid strength and avoids tarnishing. 3.05 mm diameter, 0.7 mil (18um) thickness.
Grid: Nickel
Pitch: 250 µm
Hole: 215 µm
Bar: 35 µm