Learn More
Electron Microscopy Sciences Standard Hexagonal Mesh, Material: Copper. Pitch: 500 µm. Hole: 430 µm. Bar: 70 µm., 100/PK
SDP
Catalog No. 502091171
Encompass
A reliable support specimen grid source. Features: well-defined grid bars, maximum open area, and a matte/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18 um) thickness. Material: Copper. Pitch: 500 µm . Hole: 430 µm . Bar: 70 µm.
Catalog No. 50-209-1171 Supplier Electron Microscopy Sciences Supplier No. G50HCU
A reliable support specimen grid source. Features: well-defined grid bars, maximum open area, and a matte/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18 um) thickness. Material: Copper. Pitch: 500 µm . Hole: 430 µm . Bar: 70 µm.