Learn More
Sigma Aldrich Fine Chemicals Biosciences Hexagonal weighing boats s
Catalog No. 502901427
Encompass
Bend flexible boats at points to form pour spouts. flat bottoms and sloping slides provide accurate transfer. These antistatic polystyrene boats are non-reactive to most substances. Resist temperatures to 158Af (70AC).
Catalog No. 50-290-1427 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z708577500EA
Bend flexible boats at points to form pour spouts. flat bottoms and sloping slides provide accurate transfer. These antistatic polystyrene boats are non-reactive to most substances. Resist temperatures to 158Af (70AC).