Learn More
Sigma Aldrich Fine Chemicals Biosciences Hexagonal weighing boats s
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z708615500EA
Encompass
Bend flexible boats at points to form pour spouts. flat bottoms and sloping slides provide accurate transfer. These antistatic polystyrene boats are non-reactive to most substances. Resist temperatures to 158Af (70AC).
Catalog No. 50-290-1729