Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexahydro-1,3,5-tris(2-hydroxyethyl)-s-triazine | 4719-04-4 | MFCD01678788 | >=96.0% | 219.28 g/mol | C9H21N3O3 | 500 G
SDP

Catalog No. 5000301756
Encompass_Preferred

Triazinetriethanol (hexahydro-1,3,5-tris(2-hydroxyethyl)-s-triazine) is a hexahydro triazine derivative used as a chemical reagent and intermediate. Supplied as a colorless to light-yellow liquid, it is characterized by certificate of analysis data for identity and composition.

  • CAS registry number 4719-04-4.
  • High purity: ≥96.0% (certificate of analysis).
  • Chemical formula C9H21N3O3; molecular weight 219.28 g/mol.
  • Physical form: colorless to light-yellow liquid; density ≈1.157 g/mL at 20 °C.
  • Analytical parameters reported: refractive index, pH, ethanolamine and formaldehyde content.
  • Storage recommended refrigerated or frozen for extended stability.

Catalog No. 50-003-01756 Supplier Medchemexpress LLC Supplier No. HYW018156500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triazinetriethanol (hexahydro-1,3,5-tris(2-hydroxyethyl)-s-triazine) is a hexahydro triazine derivative used as a chemical reagent and intermediate. Supplied as a colorless to light-yellow liquid, it is characterized by certificate of analysis data for identity and composition.

  • CAS registry number 4719-04-4.
  • High purity: ≥96.0% (certificate of analysis).
  • Chemical formula C9H21N3O3; molecular weight 219.28 g/mol.
  • Physical form: colorless to light-yellow liquid; density ≈1.157 g/mL at 20 °C.
  • Analytical parameters reported: refractive index, pH, ethanolamine and formaldehyde content.
  • Storage recommended refrigerated or frozen for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.